siRNA Details
Gene Symbol: SPRR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005988.2 | NP_005979.1 | 2 | 425 | ||
GenBank mRNA | CA433267.1 | - | 1 | 264 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPRR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005988.2 | NP_005979.1 | 2 | 425 | ||
GenBank mRNA | CA433267.1 | - | 1 | 264 |