siRNA Details
| Gene Symbol: SPRR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005988.2 | NP_005979.1 | 2 | 633 | ||
| GenBank mRNA | CA433267.1 | - | 1 | 56 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2A| Gene Symbol: SPRR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005988.2 | NP_005979.1 | 2 | 633 | ||
| GenBank mRNA | CA433267.1 | - | 1 | 56 | ||