siRNA Details
| Gene Symbol: STATH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009181.1 | NP_001009181.1 | 2, 3 | 165 | ||
| NM_003154.2 | NP_003145.1 | 2, 3 | 165 | |||
| GenBank mRNA | AK311812.1 | - | 2, 3 | 130 | ||
| BC067219.1 | - | 2, 3 | 131 | 30328926 | ||
| CB958641.1 | - | 2, 3 | 132 | |||
| CB986973.1 | - | 169 | ||||
| M18078.1 | - | 2, 3 | 127 | |||
| M18371.1 | - | 2, 3 | 111 | |||

STATH
Genomic Map