siRNA Details
| Gene Symbol: SULT2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004605.2 | NP_004596.2 | 3 | 619 | ||
| NM_177973.1 | NP_814444.1 | 4 | 566 | |||
| GenBank mRNA | BC034694.1 | - | 4 | 555 | 4745903 | |
| U92314.1 | - | 3 | 815 | |||
| U92315.1 | - | 4 | 566 | |||

SULT2B1
Genomic Map