siRNA Details
| Gene Symbol: SURF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278928.1 | NP_001265857.1 | 6 | 790 | ||
| NM_017503.4 | NP_059973.4 | 6 | 793 | |||
| GenBank mRNA | AI808831.1 | - | 1 | 55 | ||
| BC014411.1 | - | 6 | 762 | 4548425 | ||
| BQ672017.1 | - | 4 | 440 | |||
| CR456753.1 | - | 6 | 745 | |||
| Z35094.1 | - | 6 | 762 | |||

SURF2
Genomic Map