siRNA Details
| Gene Symbol: TBXA2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001060.5 | NP_001051.1 | 3 | 1251 | ||
| NM_201636.2 | NP_963998.2 | 3 | 1251 | |||
| XM_011528214.2 | XP_011526516.1 | 4 | 3285 | |||
| GenBank mRNA | AY429110.1 | - | 2 | 864 | ||
| BC074749.2 | - | 2 | 903 | 30915459 | ||
| BC074750.2 | - | 2 | 903 | 30915209 | ||
| BC121148.1 | - | 2 | 509 | 40121265 | ||
| U11271.1 | - | 3 | 1076 | |||
| U27325.1 | - | 3 | 987 | |||
More Information
Set Membership: |
|

TBXA2R
Genomic Map