siRNA Details
| Gene Symbol: TCEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204857.1 | NP_001191786.1 | 4 | 521 | ||
| NM_001204858.1 | NP_001191787.1 | 5 | 832 | |||
| NM_001204859.1 | NP_001191788.1 | 5 | 828 | |||
| NM_001204860.1 | NP_001191789.1 | 5 | 660 | |||
| NM_001204861.1 | NP_001191790.1 | 6 | 712 | |||
| NM_001204862.1 | NP_001191791.1 | 5 | 573 | |||
| NM_001204863.1 | NP_001191792.1 | 3 | 635 | |||
| NM_001204864.1 | NP_001191793.1 | 3 | 467 | |||
| NM_005648.3 | NP_005639.1 | 4 | 689 | |||
| XM_011517580.2 | XP_011515882.1 | 4 | 587 | |||
| XM_011517581.2 | XP_011515883.1 | 4 | 597 | |||
| GenBank mRNA | AK057889.1 | - | 4 | 569 | ||
| AL832244.1 | - | 4 | 522 | |||
| BC013809.1 | - | 4 | 513 | 3923736 | ||
| BC100283.1 | - | 4 | 649 | 6061521 | ||
| BG428202.1 | - | 4 | 523 | |||
| BX649138.1 | - | 3 | 519 | |||
| CF129635.1 | - | 3 | 454 | |||
| CX166168.1 | - | 5 | 557 | |||
| CX166843.1 | - | 4 | 628 | |||

TCEB1
Genomic Map