siRNA Details
| Gene Symbol: CLEC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003278.2 | NP_003269.2 | 1, 2 | 206 | ||
| GenBank mRNA | AK312093.1 | - | 1, 2 | 214 | ||
| BC011024.1 | - | 1, 2 | 171 | 4042921 | ||
| BI837909.1 | - | 1, 2 | 290 | |||
| CD616729.1 | - | 1, 2 | 180 | |||
| CR541981.1 | - | 1, 2 | 110 | |||
| CR542009.1 | - | 1, 2 | 110 | |||
| X64559.1 | - | 1, 2 | 203 | |||

CLEC3B
Genomic Map