siRNA Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021137.4 | NP_066960.1 | 6 | 1111 | ||
| XM_017024993.1 | XP_016880482.1 | 6 | 1067 | |||
| GenBank mRNA | AK300584.1 | - | 5 | 553 | ||
| AK312387.1 | - | 6 | 872 | |||
| BC001643.2 | - | 6 | 858 | 2823154 | ||
| BC001949.1 | - | 6 | 858 | 2823154 | ||
| BC003694.2 | - | 6 | 873 | |||
| BT020121.1 | - | 5 | 662 | |||
| CR456819.1 | - | 5 | 662 | |||
| M80783.1 | - | 6 | 815 | |||

TNFAIP1
Genomic Map