siRNA Details
| Gene Symbol: TNNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126132.2 | NP_001119604.1 | 10, 11 | 712 | ||
| NM_001126133.2 | NP_001119605.1 | 9, 10 | 679 | |||
| NM_001291774.1 | NP_001278703.1 | 10, 11 | 774 | |||
| NM_003283.5 | NP_003274.3 | 10, 11 | 712 | |||
| XM_011527246.2 | XP_011525548.1 | 9, 10 | 589 | |||
| XM_017027186.1 | XP_016882675.1 | 11, 12 | 785 | |||
| XM_017027187.1 | XP_016882676.1 | 10, 11 | 773 | |||
| GenBank mRNA | AK291352.1 | - | 10, 11 | 564 | ||
| AK308199.1 | - | 9, 10 | 531 | |||
| AY762903.1 | - | 9, 10 | 507 | |||
| AY762904.1 | - | 8, 9 | 474 | |||
| BC010963.1 | - | 10, 11 | 565 | 4291828 | ||
| BC022086.2 | - | 10, 11 | 613 | |||
| BC034143.1 | - | 10, 11 | 564 | 4328089 | ||
| BC107798.1 | - | 9, 10 | 534 | 4247379 | ||
| BT019630.1 | - | 8, 9 | 474 | |||
| M19308.1 | - | 9, 10 | 532 | |||
| M19309.1 | - | 10, 11 | 564 | |||
| S69208.1 | - | 10, 11 | 548 | |||
| S69209.1 | - | 9, 10 | 515 | |||

TNNT1
Genomic Map