siRNA Details
| Gene Symbol: TSPYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003309.3 | NP_003300.1 | 1 | 1586 | ||
| GenBank mRNA | AF042181.1 | - | 1 | 209 | ||
| AK026849.1 | - | 1 | 447 | |||
| AL136629.1 | - | 1 | 1400 | |||
| BC048969.1 | - | 1 | 1402 | 5297724 | ||
| BC068554.1 | - | 1 | 1399 | |||
| CR533535.1 | - | 1 | 1302 | |||

TSPYL1
Genomic Map