siRNA Details
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003357.4 | NP_003348.1 | 2 | 143 | ||
| GenBank mRNA | AK312165.1 | - | 2 | 143 | ||
| BC004481.2 | - | 2 | 127 | 3688615 | ||
| CR541780.1 | - | 2 | 82 | |||
| KU178373.1 | - | 2 | 82 | |||
| KU178374.1 | - | 2 | 231 | |||
| U01101.1 | - | 2 | 91 | |||
| X13197.1 | - | 2 | 89 | |||

SCGB1A1
Genomic Map