siRNA Details
| Gene Symbol: VEGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243733.1 | NP_001230662.1 | 3, 4 | 553 | ||
| NM_003377.4 | NP_003368.1 | 3, 4 | 553 | |||
| GenBank mRNA | BC008818.2 | - | 3, 4 | 343 | 4053976 | |
| U43368.1 | - | 3, 4 | 305 | |||
| U43369.1 | - | 3, 4 | 303 | |||
| U48801.1 | - | 3, 4 | 306 | |||
| U52819.1 | - | 3, 4 | 307 | |||

VEGFB
Genomic Map