siRNA Details
| Gene Symbol: VEGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003377.4 | NP_003368.1 | 6 | 743 | ||
| GenBank mRNA | BC008818.2 | - | 6 | 533 | 4053976 | |
| U43368.1 | - | 6 | 495 | |||
| U52819.1 | - | 6 | 497 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 VEGFB
VEGFB| Gene Symbol: VEGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003377.4 | NP_003368.1 | 6 | 743 | ||
| GenBank mRNA | BC008818.2 | - | 6 | 533 | 4053976 | |
| U43368.1 | - | 6 | 495 | |||
| U52819.1 | - | 6 | 497 | |||
 Back To Top
 Back To Top