siRNA Details
| Gene Symbol: MZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003422.2 | NP_003413.2 | 6 | 1109 | ||
| NM_198055.1 | NP_932172.1 | 6 | 1427 | |||
| XM_005259204.3 | XP_005259261.1 | 7 | 1782 | |||
| XM_011527264.2 | XP_011525566.1 | 6 | 1511 | |||
| XM_017027206.1 | XP_016882695.1 | 6 | 1630 | |||
| GenBank mRNA | AF055077.1 | - | 6 | 1109 | ||
| AF055078.1 | - | 6 | 1427 | |||
| BC007777.2 | - | 6 | 1085 | 4301723 | ||
| M58297.1 | - | 1 | 1209 | |||

MZF1
Genomic Map