siRNA Details
| Gene Symbol: ZNF69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321163.1 | NP_001308092.1 | 4 | 495 | ||
| NM_021915.3 | NP_068734.1 | 4 | 453 | |||
| XM_006722873.3 | XP_006722936.1 | 4 | 515 | |||
| XM_006722874.3 | XP_006722937.1 | 4 | 515 | |||
| XM_017027231.1 | XP_016882720.1 | 4 | 515 | |||
| GenBank mRNA | BC051309.1 | - | 4 | 382 | 5812091 | |
| BG393583.1 | - | 4 | 495 | |||
| BG429615.1 | - | 4 | 451 | |||

ZNF69
Genomic Map