siRNA Details
| Gene Symbol: ZNF85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256173.1 | NP_001243102.1 | 2, 3 | 240 | ||
| NM_003429.4 | NP_003420.2 | 3, 4 | 367 | |||
| GenBank mRNA | BP199573.1 | - | 3, 4 | 367 | ||
| DB119881.1 | - | 2, 3 | 189 | |||
| DN994243.1 | - | 3, 4 | 354 | |||
| M61866.1 | - | 3, 4 | 331 | |||
| U35376.1 | - | 3, 4 | 337 | |||

ZNF85
Genomic Map