siRNA Details
| Gene Symbol: ZNF117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015852.3 | NP_056936.2 | 3, 4 | 1310 | ||
| GenBank mRNA | BC034021.1 | - | 4, 5 | 463 | ||
| M27879.1 | - | 2, 3 | 131 | |||
| M55422.1 | - | 3, 4 | 1309 | |||
| M61867.1 | - | 2, 3 | 131 | |||

ZNF117
Genomic Map