siRNA Details
Gene Symbol: ZNF137P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023311.1 | 1 | 270 | |||
GenBank mRNA | BC110455.1 | - | 1 | 270 | ||
BC110456.1 | - | 1 | 270 | |||
U09414.1 | - | 1 | 304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF137P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023311.1 | 1 | 270 | |||
GenBank mRNA | BC110455.1 | - | 1 | 270 | ||
BC110456.1 | - | 1 | 270 | |||
U09414.1 | - | 1 | 304 |