siRNA Details
| Gene Symbol: SLC30A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004434.2 | NP_001004434.1 | 7 | 1096 | ||
| NM_032513.4 | NP_115902.1 | 6 | 949 | |||
| GenBank mRNA | AF370409.1 | - | 7 | 943 | ||
| AK023491.1 | - | 2 | 118 | |||
| AK023504.1 | - | 2 | 38 | |||
| BC006251.2 | - | 6 | 924 | 3948165 | ||

SLC30A2
Genomic Map