siRNA Details
| Gene Symbol: HIST3H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003493.2 | NP_003484.1 | 1 | 20 | ||
| GenBank mRNA | AK312217.1 | - | 1 | 53 | ||
| BC069079.1 | - | 1 | 20 | 7216893 | ||
| BC101837.1 | - | 1 | 68 | 8069343 | ||
| BC101839.1 | - | 1 | 68 | 8069345 | ||
| CR542013.1 | - | 1 | 20 | |||

HIST3H3
Genomic Map