siRNA Details
| Gene Symbol: HIST1H2AC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003512.3 | NP_003503.1 | 1 | 364 | ||
| GenBank mRNA | AK311811.1 | - | 1 | 337 | ||
| BC017379.1 | - | 1 | 328 | |||
| BC050602.1 | - | 1 | 320 | |||
| BC085010.1 | - | 1 | 325 | |||
| U90551.1 | - | 1 | 373 | |||

HIST1H2AC
Genomic Map