siRNA Details
| Gene Symbol: HIST1H3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003537.3 | NP_003528.1 | 1 | 16 | ||
| GenBank mRNA | BC069133.1 | - | 1 | 16 | 7216989 | |
| BC096132.1 | - | 1 | 16 | 40002522 | ||
| BC096133.3 | - | 1 | 16 | 40002523 | ||
| BC096134.3 | - | 1 | 16 | 40002524 | ||
| BC099630.3 | - | 1 | 19 | 40002521 | ||

HIST1H3B
Genomic Map