siRNA Details
| Gene Symbol: HIST1H4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003547.2 | NP_003538.1 | 1 | 267 | ||
| GenBank mRNA | BC126276.1 | - | 1 | 321 | 8991992 | |
| BC126278.1 | - | 1 | 321 | 8991994 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H4G| Gene Symbol: HIST1H4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003547.2 | NP_003538.1 | 1 | 267 | ||
| GenBank mRNA | BC126276.1 | - | 1 | 321 | 8991992 | |
| BC126278.1 | - | 1 | 321 | 8991994 | ||