siRNA Details
| Gene Symbol: CST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003650.3 | NP_003641.3 | 3 | 618 | ||
| GenBank mRNA | AB015225.1 | - | 3 | 490 | ||
| AF031824.1 | - | 3 | 557 | |||
| AF036342.1 | - | 3 | 399 | |||
| BC015507.1 | - | 3 | 513 | 3893020 | ||
| BT009825.1 | - | 3 | 408 | |||
| CA449286.1 | - | 2 | 319 | |||
| CR541860.1 | - | 3 | 342 | |||
| CR541878.1 | - | 3 | 342 | |||

CST7
Genomic Map