siRNA Details
| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075140.1 | - | 5 | 387 | ||
| AK309540.1 | - | 6 | 416 | |||
| AY358081.1 | - | 5 | 376 | |||
| AY358283.1 | - | 5 | 413 | |||
| BC020731.1 | - | 5 | 387 | 4692272 | ||
| BG546674.1 | - | 6 | 448 | |||
| CR456808.1 | - | 6 | 414 | |||

FCN3
Genomic Map