siRNA Details
| Gene Symbol: KRT37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003770.4 | NP_003761.3 | 7 | 1417 | ||
| GenBank mRNA | AJ786655.1 | - | 7 | 1253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT37| Gene Symbol: KRT37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003770.4 | NP_003761.3 | 7 | 1417 | ||
| GenBank mRNA | AJ786655.1 | - | 7 | 1253 | ||