siRNA Details
| Gene Symbol: S1PR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003775.3 | NP_003766.1 | 1 | 882 | ||
| GenBank mRNA | AJ000479.1 | - | 1 | 849 | ||
| BC014970.1 | - | 1 | 852 | 4849349 | ||
| FJ200495.1 | - | 2 | 689 | |||
More Information
Set Membership: |
|

S1PR4
Genomic Map