siRNA Details
Gene Symbol: ADAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002224.2 | 2 | 1709 | |||
GenBank mRNA | AA609214.1 | - | 1 | 139 | ||
AI024595.1 | - | 1 | 133 | |||
AK093400.1 | - | 2 | 1709 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002224.2 | 2 | 1709 | |||
GenBank mRNA | AA609214.1 | - | 1 | 139 | ||
AI024595.1 | - | 1 | 133 | |||
AK093400.1 | - | 2 | 1709 |