siRNA Details
| Gene Symbol: HIST1H2AG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC016677.1 | - | 1 | 2137 | 4342191 | |
| L19778.1 | - | 1 | 2148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AG| Gene Symbol: HIST1H2AG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC016677.1 | - | 1 | 2137 | 4342191 | |
| L19778.1 | - | 1 | 2148 | |||