siRNA Details
| Gene Symbol: HIST1H2AG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021064.4 | NP_066408.1 | 1 | 80 | ||
| GenBank mRNA | AK312124.1 | - | 1 | 80 | ||
| BC016677.1 | - | 1 | 65 | 4342191 | ||
| L19778.1 | - | 1 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AG| Gene Symbol: HIST1H2AG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021064.4 | NP_066408.1 | 1 | 80 | ||
| GenBank mRNA | AK312124.1 | - | 1 | 80 | ||
| BC016677.1 | - | 1 | 65 | 4342191 | ||
| L19778.1 | - | 1 | 76 | |||