siRNA Details
| Gene Symbol: HIST1H2BJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021058.3 | NP_066402.2 | 1 | 382 | ||
| GenBank mRNA | AK311849.1 | - | 1 | 410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2BJ| Gene Symbol: HIST1H2BJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021058.3 | NP_066402.2 | 1 | 382 | ||
| GenBank mRNA | AK311849.1 | - | 1 | 410 | ||