siRNA Details
| Gene Symbol: H1FX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006026.3 | NP_006017.1 | 1 | 678 | ||
| GenBank mRNA | BC000426.2 | - | 1 | 678 | 2819756 | |
| BC010435.1 | - | 1 | 384 | 3538400 | ||
| D64142.1 | - | 1 | 404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H1FX| Gene Symbol: H1FX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006026.3 | NP_006017.1 | 1 | 678 | ||
| GenBank mRNA | BC000426.2 | - | 1 | 678 | 2819756 | |
| BC010435.1 | - | 1 | 384 | 3538400 | ||
| D64142.1 | - | 1 | 404 | |||