siRNA Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286526.1 | NP_001273455.1 | 2 | 476 | ||
| NM_004765.3 | NP_004756.2 | 2 | 476 | |||
| XM_011545980.2 | XP_011544282.1 | 2 | 724 | |||
| XM_017023885.1 | XP_016879374.1 | 2 | 717 | |||
| XM_017023886.1 | XP_016879375.1 | 2 | 722 | |||
| XM_017023887.1 | XP_016879376.1 | 2 | 722 | |||
| XM_017023888.1 | XP_016879377.1 | 2 | 722 | |||
| GenBank mRNA | AJ223980.1 | - | 2 | 118 | ||
| BC019071.1 | - | 2 | 252 | 4909589 | ||
| BC058863.1 | - | 2 | 476 | 5167136 | ||

BCL7C
Genomic Map