siRNA Details
| Gene Symbol: CPNE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280558.1 | NP_001267487.1 | 16 | 1664 | ||
| NM_006032.3 | NP_006023.1 | 15 | 1652 | |||
| XM_005268216.1 | XP_005268273.1 | 16 | 1608 | |||
| XM_005268217.1 | XP_005268274.1 | 14 | 1302 | |||
| GenBank mRNA | AB009288.1 | - | 15 | 1452 | ||
| AK125195.1 | - | 14 | 2214 | |||
| AK222675.1 | - | 15 | 1412 | |||
| AK293655.1 | - | 16 | 1664 | |||
| AK314182.1 | - | 15 | 1465 | |||
| AL050397.1 | - | 3 | 1315 | |||
| AL137688.1 | - | 7 | 3980 | |||
| BC018046.1 | - | 15 | 1458 | 4799650 | ||

CPNE6
Genomic Map