siRNA Details
| Gene Symbol: CSAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129826.2 | NP_001123298.1 | 1 | 229 | ||
| NM_001129828.2 | NP_001123300.1 | 1 | 229 | |||
| GenBank mRNA | AF080246.1 | - | 1 | 229 | ||
| AF136715.1 | - | 1 | 229 | |||
| BC119734.1 | - | 1 | 202 | 40119148 | ||
| BC119735.1 | - | 1 | 201 | 40119150 | ||
| Gene Symbol: CSAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724857.3 | XP_006724920.1 | 1 | 355 | ||
| XM_017030023.1 | XP_016885512.1 | 1 | 355 | |||

CSAG3
Genomic Map