siRNA Details
| Gene Symbol: CSAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF080246.1 | - | 1 | 313 | ||
| AF136715.1 | - | 1 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSAG3| Gene Symbol: CSAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF080246.1 | - | 1 | 313 | ||
| AF136715.1 | - | 1 | 313 | |||