siRNA Details
Gene Symbol: VPS9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004913.2 | NP_004904.2 | 5 | 600 | ||
XM_005256329.4 | XP_005256386.1 | 5 | 660 | |||
XM_005256330.4 | XP_005256387.1 | 5 | 477 | |||
XM_005256331.2 | XP_005256388.1 | 2 | 134 | |||
XM_006721350.2 | XP_006721413.1 | 5 | 660 | |||
XM_011523476.2 | XP_011521778.1 | 5 | 657 | |||
XM_011523477.2 | XP_011521779.1 | 4 | 2473 | |||
XM_011523478.2 | XP_011521780.1 | 4 | 2470 | |||
XM_011523479.2 | XP_011521781.1 | 5 | 660 | |||
XM_011523480.2 | XP_011521782.1 | 5 | 514 | |||
GenBank mRNA | AB018551.1 | - | 4 | 641 | ||
BY795348.2 | - | 2 | 221 |
