siRNA Details
| Gene Symbol: KIAA0100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014680.3 | NP_055495.2 | 11 | 1300 | ||
| XM_005258073.2 | XP_005258130.1 | 11 | 1295 | |||
| XM_017025423.1 | XP_016880912.1 | 10 | 1149 | |||
| GenBank mRNA | AK130007.1 | - | 11 | 1255 | ||
| BC048096.1 | - | 9 | 1808 | |||
| BC050440.1 | - | 11 | 1296 | |||
| BC120873.1 | - | 11 | 1159 | 40075102 | ||
| CR933634.1 | - | 11 | 1252 | |||
| D43947.3 | - | 11 | 1272 | |||

KIAA0100
Genomic Map