siRNA Details
| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 6 | 3369 | |||
| GenBank mRNA | BC152473.1 | - | 1 | 1648 | ||
| D50915.1 | - | 1 | 1648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0125| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 6 | 3369 | |||
| GenBank mRNA | BC152473.1 | - | 1 | 1648 | ||
| D50915.1 | - | 1 | 1648 | |||