siRNA Details
| Gene Symbol: TRANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014831.2 | NP_055646.2 | 21 | 8125 | ||
| XM_006713432.3 | XP_006713495.1 | 22 | 8293 | |||
| XM_011534289.2 | XP_011532591.1 | 22 | 9054 | |||
| XM_017007570.1 | XP_016863059.1 | 21 | 7925 | |||
| XM_017007571.1 | XP_016863060.1 | 22 | 8186 | |||
| XM_017007572.1 | XP_016863061.1 | 22 | 8088 | |||
| GenBank mRNA | AB002340.3 | - | 11 | 6503 | ||

TRANK1
Genomic Map