siRNA Details
| Gene Symbol: TBC1D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286658.2 | NP_001273587.1 | 1 | 862 | ||
| NM_001286659.2 | NP_001273588.1 | 1 | 862 | |||
| NM_014832.4 | NP_055647.2 | 1 | 862 | |||
| XM_005266603.1 | XP_005266660.1 | 1 | 1340 | |||
| XM_011535331.1 | XP_011533633.1 | 1 | 1340 | |||
| GenBank mRNA | AB011175.1 | - | 1 | 808 | ||
| AB449885.1 | - | 1 | 461 | |||
| AK307493.1 | - | 1 | 862 | |||
| BC151239.1 | - | 1 | 800 | 8860512 | ||
| FM207106.1 | - | 1 | 461 | |||
| FM207107.1 | - | 1 | 461 | |||

TBC1D4
Genomic Map