siRNA Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242925.1 | NP_001229854.1 | 2 | 217 | ||
| NM_014858.3 | NP_055673.2 | 2 | 881 | |||
| XM_005245684.1 | XP_005245741.1 | 2 | 572 | |||
| XM_005245685.4 | XP_005245742.1 | 2 | 203 | |||
| XM_005245686.3 | XP_005245743.1 | 2 | 598 | |||
| GenBank mRNA | AB007950.3 | - | 2 | 881 | ||
| AK131419.1 | - | 3 | 670 | |||
| AK293734.1 | - | 2 | 217 | |||
| BC131629.1 | - | 2 | 353 | 40118729 | ||
| DC309246.1 | - | 2 | 215 | |||

TMCC2
Genomic Map