siRNA Details
| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006041.2 | NP_006032.1 | 1 | 494 | ||
| NR_130138.1 | 1 | 494 | ||||
| GenBank mRNA | AF105377.1 | - | 1 | 355 | ||
| AK023723.1 | - | 1 | 461 | |||
| BC063301.1 | - | 1 | 411 | 30343352 | ||
| BC069664.1 | - | 1 | 295 | 7262483 | ||
| BC069725.1 | - | 1 | 294 | 7262495 | ||
| DA388077.1 | - | 1 | 495 | |||

HS3ST3B1
Genomic Map