siRNA Details
| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006042.2 | NP_006033.1 | 2 | 2018 | ||
| XM_011524114.2 | XP_011522416.1 | 3 | 2262 | |||
| GenBank mRNA | AF105376.1 | - | 2 | 2003 | ||
| AK292047.1 | - | 2 | 2017 | |||
| AY358838.1 | - | 2 | 1977 | |||
| BC044647.1 | - | 2 | 1368 | 5478042 | ||

HS3ST3A1
Genomic Map