siRNA Details
| Gene Symbol: RCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032279.1 | NP_001027450.1 | 1 | 149 | ||
| NM_005133.2 | NP_005124.1 | 1 | 149 | |||
| GenBank mRNA | AF121951.1 | - | 1 | 149 | ||
| AL713740.1 | - | 1 | 726 | |||
| BC052622.1 | - | 1 | 110 | |||
| BC093726.1 | - | 1 | 145 | 7939571 | ||
| BC093728.1 | - | 1 | 144 | 7939573 | ||
| Y13835.2 | - | 1 | 118 | |||

RCE1
Genomic Map