siRNA Details
| Gene Symbol: FRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005479.3 | NP_005470.2 | 1 | 802 | ||
| GenBank mRNA | AB074890.1 | - | 1 | 726 | ||
| AK307245.1 | - | 2 | 481 | |||
| BC034476.1 | - | 1 | 781 | 4823463 | ||
| U58975.2 | - | 1 | 802 | |||

FRAT1
Genomic Map