siRNA Details
| Gene Symbol: SCAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005698.3 | NP_005689.2 | 3 | 473 | ||
| NM_052837.2 | NP_443069.1 | 2 | 395 | |||
| XM_006711105.3 | XP_006711168.1 | 3 | 791 | |||
| XM_016999991.1 | XP_016855480.1 | 3 | 791 | |||
| GenBank mRNA | AF005039.1 | - | 3 | 335 | ||
| BC000161.2 | - | 3 | 347 | 2901296 | ||
| BC005135.2 | - | 3 | 320 | 3510027 | ||
| BC010505.1 | - | 2 | 264 | 3452347 | ||
| BM790077.1 | - | 2 | 369 | |||
| BU193466.1 | - | 2 | 395 | |||
| CR541757.1 | - | 3 | 239 | |||

SCAMP3
Genomic Map