siRNA Details
| Gene Symbol: PREB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013388.4 | NP_037520.1 | 8 | 1338 | ||
| XM_006711914.2 | XP_006711977.1 | 7 | 1145 | |||
| GenBank mRNA | AF203687.1 | - | 8 | 1216 | ||
| AF226684.1 | - | 8 | 1229 | |||
| AK023064.1 | - | 8 | 1295 | |||
| AK023405.1 | - | 8 | 1209 | |||
| AK292178.1 | - | 9 | 1225 | |||
| BC002765.2 | - | 8 | 1269 | 3631721 | ||
| BC012890.1 | - | 8 | 1279 | 4158577 | ||
| BC016472.1 | - | 8 | 1269 | 4178911 | ||
| BC016906.1 | - | 8 | 1212 | 4427483 | ||
| BC041032.1 | - | 8 | 1332 | 4638313 | ||

PREB
Genomic Map