siRNA Details
| Gene Symbol: SSX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021014.3 | NP_066294.1 | 4 | 362 | ||
| XM_011543885.2 | XP_011542187.1 | 4 | 315 | |||
| GenBank mRNA | AK310348.1 | - | 4 | 304 | ||
| AK310513.1 | - | 4 | 315 | |||
| BC005904.1 | - | 4 | 316 | 4064072 | ||
| BC103862.1 | - | 4 | 361 | 40003336 | ||
| DB444418.1 | - | 4 | 423 | |||
| S82471.1 | - | 3 | 222 | |||
| U90840.1 | - | 4 | 275 | |||

SSX3
Genomic Map